missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L1/PGP9.5 Antibody (rUCHL1/775), Novus Biologicals™
Mouse Monoclonal Antibody
290.00 € - 509.00 €
Specifications
| Antigen | UCH-L1/PGP9.5 |
|---|---|
| Clone | rUCHL1/775 |
| Concentration | 0.2 mg/mL |
| Dilution | Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array |
| Applications | Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18681948
|
Novus Biologicals
NBP2-75773-20ug |
20 μg |
290.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672547
|
Novus Biologicals
NBP2-75773-100ug |
100 μg |
509.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCH-L1/PGP9.5 Monoclonal antibody specifically detects UCH-L1/PGP9.5 in Human, Rat, Canine samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Protein ArraySpecifications
| UCH-L1/PGP9.5 | |
| 0.2 mg/mL | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Alzheimers Research, Cellular Markers, Neurodegeneration, Neuronal Cell Markers, Neuroscience, Neurotransmission | |
| 10 mM PBS with 0.05% BSA | |
| EC 3.4.19.12, EC 6.-, Neuron cytoplasmic protein 9.5, PARK5, PGP 9.5, PGP9.5, PGP9.5Uch-L1, PGP95, ubiquitin carboxyl-terminal esterase L1 (ubiquitin thiolesterase), ubiquitin carboxyl-terminal hydrolase isozyme L1, ubiquitin C-terminal hydrolase, Ubiquitin thioesterase L1, UCHL1, UCH-L1 | |
| Recombinant full-length human UCH-L1/PGP9.5 protein (Uniprot: P09936) | |
| Primary | |
| Store at 4°C. |
| rUCHL1/775 | |
| Western Blot 1-2 μg/mL, Flow Cytometry 1-2 μg/million cells, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence 1-2 μg/mL, Immunohistochemistry-Paraffin 1-2 μg/mL, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Canine | |
| P09936 | |
| 7345 | |
| IgG1 κ | |
| Protein A or G purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel