missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCH-L5/UCH37 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
201.00 € - 470.00 €
Specifications
| Antigen | UCH-L5/UCH37 |
|---|---|
| Dilution | Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18699650
|
Novus Biologicals
NBP2-93687-0.02ml |
0.02 mL |
201.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18638511
|
Novus Biologicals
NBP2-93687-0.1ml |
0.1 mL |
470.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UCH-L5/UCH37 Polyclonal antibody specifically detects UCH-L5/UCH37 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| UCH-L5/UCH37 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cell Biology | |
| PBS (pH 7.3), 50% glycerol | |
| 51377 | |
| IgG | |
| Affinity purified |
| Western Blot 1:100 - 1:500, Immunohistochemistry 1:50 - 1:200, Immunocytochemistry/ Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| EC 3.4.19.12, INO80 complex subunit R, INO80R, ubiquitin carboxyl-terminal esterase L5, ubiquitin carboxyl-terminal hydrolase isozyme L5, ubiquitin carboxyl-terminal hydrolase L5, Ubiquitin C-terminal hydrolase UCH37, Ubiquitin thioesterase L5, UCH37CGI-70, UCH-L5 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-328 of human UCHL5 (NP_001186190.1). MTGNAGEWCLMESDPGVFTELIKGFGCRGAQVEEIWSLEPENFEKLKPVHGLIFLFKWQPGEEPAGSVVQDSRLDTIFFAKQVINNACATQAIVSVLLNCTHQDVHLGETLSEFKEFSQSFDAAMKGLALSNSDVIRQVHNSFARQQMFEFDTKTSAKEEDAFHFVSYVPVNGRLYELDGLREGPIDLGACNQDDWISAVRPVIEKRIQKYSEGEIRFNLMAIVSDRKMIYEQKIAELQRQLAEEPMDTDQGNSMLSAIQSEVAKNQMLIEEEVQKLKRYKIENIRRKHNYLPFIMELLKTLAEHQQLIPLVEKAKEKQNAKKAQETK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title