Learn More
UCHL5 Mouse, anti-Human, Clone: 2B8G11, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 68535-1-IG-20UL
This item is not returnable.
View return policy
Description
The UCHL5 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human UCHL5. This antibody recognizes human antigen. The UCHL5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| UCHL5 | |
| 2B8G11 | |
| CGI 70, INO80R, Ubiquitin thioesterase L5, UCH L5, UCH37, UCHL5 | |
| 20 μL | |
| 51377 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
| Western Blot, ELISA | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human | |
| 150 kDa | |
| IgG2a |