missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP1 Antibody (Vab12 P4B12*A12) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50343-0.025ml
This item is not returnable.
View return policy
Description
UCP1 Monoclonal antibody specifically detects UCP1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry
Specifications
| UCP1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| SLC25A7mitochondrial brown fat uncoupling protein 1, Solute carrier family 25 member 7, thermogenin, UCP, UCP 1, uncoupling protein 1 (mitochondrial, proton carrier) | |
| Ovalbumin-conjugated synthetic peptide - (10 amino acids) derived from the middle polypeptide region of human UCP1. The exact sequence is proprietary. | |
| 0.025 mL | |
| Cancer, Cellular Signaling, metabolism | |
| 7350 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 κ |
| Western Blot, ELISA, Immunohistochemistry | |
| Vab12 P4B12*A12 | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction