missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UCP3 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94663-0.02ml
This item is not returnable.
View return policy
Description
UCP3 Polyclonal antibody specifically detects UCP3 in Human, Mouse samples. It is validated for Western Blot
Specifications
| UCP3 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| mitochondrial uncoupling protein 3, SLC25A9Solute carrier family 25 member 9, UCP 3, uncoupling protein 3 (mitochondrial, proton carrier), Uncoupling protein-3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-110 of human UCP3 (NP_073714.1). MVGLKPSDVPPTMAVKFLGAGTAACFADLVTFPLDTAKVRLQIQGENQAVQTARLVQYRGVLGTILTMVRTEGPCSPYNGLVAGLQRQMSFASIRIGLYDSVKQVYTPKG | |
| 0.02 mL | |
| Cancer, Cardiovascular Biology, Diabetes Research, Endocrinology, metabolism, Neuroscience, Signal Transduction | |
| 7352 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction