missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ UIP1 Antibody (SR4268), Novus Biologicals™
Rabbit Monoclonal Antibody
316.00 € - 382.00 €
Specifications
| Antigen | UIP1 |
|---|---|
| Clone | SR4268 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Preis | Menge & Verfügbarkeit | |||||
|
30041574
|
Novus Biologicals™
NBP3-27829-100ul |
100 μL |
382.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041679
|
Novus Biologicals™
NBP3-27829-50ul |
50 μL |
316.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
UIP1 Monoclonal antibody specifically detects UIP1 in Human samples. It is validated for Western BlotSpezifikation
| UIP1 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 55559 | |
| IgG | |
| Affinity purified |
| SR4268 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| 26S proteasome-associated UCH37-interacting protein 1, expressed-Xq28STS protein, HAUS augmin-like complex subunit 7, HAUS augmin-like complex, subunit 7, UCH37 interacting protein 1, UCHL5 interacting protein, UCHL5-interacting protein, UCHL5IPHSXQ28ORF, UIP126S proteasome-associated UCH interacting protein 1, X-linked protein STS1769 | |
| A synthesized peptide derived from human UIP1 (Uniprot #: Q99871) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts