missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ULBP-2/5/6 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 43 publications
241.00 € - 2155.00 €
Specifications
| Antigen | ULBP-2/5/6 |
|---|---|
| Clone | 165903 |
| Concentration | LYOPH |
| Dilution | Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1 ug/mL, CyTOF-ready |
| Applications | Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18751728
![]() |
R&D Systems
MAB1298-SP |
25 μg |
241.00 €
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18744487
![]() |
R&D Systems
MAB1298-100 |
100 μg |
533.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18733027
![]() |
R&D Systems
MAB1298-500 |
500 μg |
2155.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ULBP-2/5/6 Monoclonal specifically detects ULBP-2/5/6 in Human samples. It is validated for Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF-ready.Specifications
| ULBP-2/5/6 | |
| LYOPH | |
| Flow Cytometry, Blockade of Receptor-ligand Interaction, CyTOF | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9BZM5 | |
| 80328 | |
| IgG2a | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 165903 | |
| Flow Cytometry 0.25 ug/10^6 cells, Blockade of Receptor-ligand Interaction 1 ug/mL, CyTOF-ready | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ALCAN-alpha, N2DL2, NKG2DL2, RAET1G, RAET1H, RAET1L, UL16 binding protein 2, ULBP2, ULBP5, ULBP6 | |
| BaF3 mouse pro-B cell line transfected with human ULBP-2 Accession # Q9BZM5 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human ULBP-2 and human RAET1L/ULBP-6 in direct ELISA and stains cells transfected with human ULBP-2, human ULBP-5 or human RAET1L/ULBP-6 in flow cytometry. In direct ELISA, 11% cross-reactivity with recombinant human ULBP5 is observed. It does not stain cells transfected with human ULBP-1 or human ULBP-3 in flow cytometry. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title