missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Ulex Europaeus Lectin 1 Native Protein

Product Code. 18695845 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18695845 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18695845 Supplier R&D Systems™ Supplier No. NBP2427005mg
  • ONLINE EXCLUSIVE PRICE
    24.55 €
    21.00 € / 5mg
    Save 3.55 €
    14% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Native Protein

Agglutination activity is expressed in μg/mL and is determined from serial dilutions in phosphate buffered saline, pH 6.8, of a 1 mg/mL solution. This activity is the lowest concentration to agglutinate a 2% suspension of human blood group O erythrocytes after 1 hour incubation at 25 degrees C.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) SDS-PAGE, Bioactivity
Formulation Lyophilized in buffer. Information about buffer components is properietary.
Molecular Weight (g/mol) MolecularWeight-theroretical: 68 kDa
Quantity 5 mg
Storage Requirements Store at 4°C. Do not freeze.
Gene Alias Gorse or Furze Lectin, UEA-1; H(O) lectin 1; Anti H(O) lectin I
Conjugate Unconjugated
Species Plants
Recombinant Native
Reconstitution Reconstitute with PBS (pH 6.8) to a final concentration of 1 mg/mL
Inhibitor Properties Ulex europaeus agglutinin has anti-H blood group specificity.
Form Purified
Product Line Novus
Purity or Quality Grade >50% SDS-PAGE
Protein Ulex Europaeus Lectin 1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.