missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ULK1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 7 publications
Brand: Novus Biologicals NBP2-24738SS
This item is not returnable.
View return policy
Description
ULK1 Polyclonal specifically detects ULK1 in Human, Mouse, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ULK1 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.05% Sodium Azide | |
| ATG1, ATG1 autophagy related 1 homolog, ATG1A, EC 2.7.11, EC 2.7.11.1, FLJ38455, FLJ46475, KIAA0722, serine/threonine-protein kinase ULK1, UNC51, unc-51 (C. elegans)-like kinase 1, Unc51.1, Unc-51-like kinase 1, unc-51-like kinase 1 (C. elegans) | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human and mouse. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.0 mg/mL | |
| Western Blot 1-3 ug/ml, Immunohistochemistry, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 27740626), Immunohistochemistry-Paraffin 10 ug/ml | |
| O75385 | |
| ULK1 | |
| A synthetic peptide corresponding to amino acids between 320 and 370 of human ULK1 was used as the immunogen, GenBank no. sp O75385.2 ULK1_HUMAN. | |
| 0.025 mg | |
| Autophagy, mTOR Pathway, Protein Kinase | |
| 8408 | |
| Human, Mouse, Monkey | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction