missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Ultra Rainbow Calibration ERF Particle Set (6 peaks)
Shop All Bio Techne ProductsDescription
- Contains 6 intensities of Ultra Rainbow Fluorescent Beads with NIST assigned ERF (Equivalent Number of Reference Fluorophores) values based on a published procedure using NIST SRM 1934 and a calibrated laserbased CCD fluorimeter.
- Provides microsphere reference standards for fluorescein isothiocyanate (FITC), phycoerythrin (PE), allophycocyanin (APC) and Pacific Blue (PB) fluorescent channels using four different NIST reference fluorophores; fluorescein, Nile Red, APC and Coumarin 30.
Specifications
Specifications
| Content And Storage | Store at 4C in the dark. Do not freeze. |
| For Use With (Application) | Flow Cytometry, Fluorescence Imaging |
| Quantity | 1 Kit |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?