missing translation for 'onlineSavingsMsg'
Learn More

Merck Ultrafiltration Discs, 50 kDa NMW, Biomax Polyethersulfone

Product Code. 12711583 Shop All Merck Products
Click to view available options
Diameter (Metric) Filter:
25 mm
44.5 mm
63.5 mm
76 mm
Filtration Area:
13.4 cm2
28.7 cm2
4.1 cm2
41.8 cm2
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 12711583

Brand: Merck PBQK04310

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

50 kDa NMWL, Biomax polyethersulfone. Assemble your Amicon Stirred Cell with the membrane of your choice.

Assemble your Amicon Stirred Cell with the membrane of your choice. Ultrafiltration cut disc membranes are available in Ultracel™ regenerated cellulose and Biomax™ polyethersulvone (PES). The membranes are available in a range of different pore sizes & diameters.
Ultracel regenerated cellulose membranes are recommended for concentrating or desalting dilute solutions. The hydrophilic, tight microstructure of Ultracel membranes assures the highest possible retention with the lowest possible adsorption of protein, DNA or other macromolecules.
Biomax PES membranes are recommended for concentrating or desalting higher volumes of more concentrated samples such as serum, plasma, or conditioned tissue culture media.
Microfiltration Durapore™ PVDF cut discs are available 0.1 μm, 0.22 μm, and 0.45 μm pore sizes in both 63.5 mm and 75 mm diameters.

Features & Benefits:

  • Disc diameters include: 25, 44.5, 47, 63.5, 76, 90 and 150 mm
  • Both Ultracel™, Durapore™, and Biomax™ membranes are available in a wide range of pore sizes to meet your separation requirements
  • Compatible with Amicon™ Stirred Cells

Applications:
Concentration, Buffer Exchange, Desalting

TRUSTED_SUSTAINABILITY

Specifications

Filter Type Ultrafiltration
Format Ultrafiltration Disc
Material PES
MWCO 50 kDa
Filter Code PBQK
Filtration Area 13.4 cm2
For Use With (Application) Protein purification; protein concentration; protein desalting
Housing Material PES
Quantity 10Pack
Diameter (English) 1.75 in.
Diameter (Metric) 44.5 mm
Diameter (Metric) Filter 44.5 mm
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.