missing translation for 'onlineSavingsMsg'
Learn More

Ultramark∣r 1621, Mass Spec Std

Product Code. p-7046887
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
This item is not returnable. View return policy

Product Code. 11359335

Brand: Thermo Scientific Alfa Aesar L16698.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

Edge
This item is not returnable. View return policy

CAS: 105809-15-2 | C6H18N3P3 | 231.189 g/mol

Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Ultramark 1621 is used as a reference compound for positive and negative ion fast-atom bombardment high-resolution mass spectrometry.

Solubility
Miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105809-15-2
Molecular Formula C6H18N3P3
Molecular Weight (g/mol) 231.189
MDL Number MFCD00242454
InChI Key DEBZEVJNWCNATM-MZWXYZOWSA-N
PubChem CID 44119860
IUPAC Name 2,2,4,4,6,6-hexakis(deuteriomethyl)-1,3,5-triaza-2$l^{5},4$l^{5},6$l^{5}-triphosphacyclohexa-1,3,5-triene
SMILES CP1(=NP(=NP(=N1)(C)C)(C)C)C

Specifications

Density 1.8
Solubility Miscible with water
Quantity 250 mg
Formula Weight 2120
Chemical Name or Material Ultramark™ 1621, Mass Spec Standard

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.