missing translation for 'onlineSavingsMsg'
Learn More

Umeclidinium bromide, MedChemExpress

Product Code. 16487717
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16487717 5 mg 5mg
16464988 10 mg 10mg
16425756 100 mg 100mg
16411237 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16487717 Supplier Medchem Express Supplier No. HY12100
 Restricted Item
Estimated Shipment: 14-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM.

Umeclidinium bromide is a novel mAChR antagonist. The affinity (Ki) of Umeclidinium bromide for the cloned human M1-M5 mAChRs ranges from 0.05 to 0.16 nM.

Specifications

Chemical Name or Material Umeclidinium bromide
Physical Form Solid
CAS 869113-09-7
Color Off-White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C29H34BrNO2
For Use With (Application) COVID-19-immunoregulation
Synonym GSK573719A
Solubility Information DMSO : ≥ 34 mg/mL (66.86 mM)
SMILES OC(C12CC[N+](CC2)(CCOCC3=CC=CC=C3)CC1)(C4=CC=CC=C4)C5=CC=CC=C5.[Br-]
Molecular Weight (g/mol) 508.49
Formula Weight 508.49
Percent Purity 96.01%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.