missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ UMODL1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA598230
This item is not returnable.
View return policy
Description
UMODL1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
UMODL1 gene ontology annotations related to this gene include adipose tissue development; cellular response to gonadotropin-releasing hormone; multicellular organismal reproductive process; regulation of apoptotic process; regulation of gene expression; regulation of ovarian follicle development; single fertilization.
Specifications
| UMODL1 | |
| Polyclonal | |
| Unconjugated | |
| UMODL1 | |
| D17Ertd488e; Olfactorin; RGD1566039; Umodl1; uromodulin like 1; uromodulin-like 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 52020, 89766 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 2.56 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q5DID0, Q5DID3 | |
| UMODL1 | |
| Recombinant Human Uromodulin-like 1 protein (537-654AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction