missing translation for 'onlineSavingsMsg'
Learn More

UNC0646, MedChemExpress

Product Code. 16498517
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16498517

Brand: Medchem Express HY13807

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress UNC0646 is a potent and selective histone methyltransferase G9a inhibitor with an IC50 of 6 nM. UNC0646 is also a potent GLP inhibitor (IC50 G9a/GLP over SETD7, SUV39H2, SETD8 and PRMT3. UNC0646 reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 26 nM.

UNC0646 is a potent and selective histone methyltransferase G9a inhibitor with an IC50 of 6 nM. UNC0646 is also a potent GLP inhibitor (IC50 G9a/GLP over SETD7, SUV39H2, SETD8 and PRMT3. UNC0646 reduces H3K9me2 levels in MDA-MB-231 cells with an IC50 of 26 nM.

Specifications

Chemical Name or Material UNC0646
CAS 1320288-17-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C36H59N7O2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (80.40 mM; ultrasonic and warming and adjust pH to 3 with HCl and heat to 80°C)
SMILES COC1=CC2=C(NC3CCN(C4CCCCC4)CC3)N=C(N5CCN(C(C)C)CCC5)N=C2C=C1OCCCN6CCCCC6
Molecular Weight (g/mol) 621.9
Formula Weight 621.9
Percent Purity 99.13%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.