Learn More
Scientific Industries Universal Clip Plates
Product Code. 10790204 Shop All Ohaus Products
Change view
Click to view available options
Packaging Quantity:
2 Clip Plates
Unit Size:
Pack of 2
Product Code. Packaging Quantity Unit Size
10790204 2 Clip Plates Pack of 2
1 options
Product Code. 10790204 Supplier Scientific Industries Supplier No. 83041374
Estimated Shipment: 07-09-2026
Add to basket
Add to basket
Universal Clip Plates are accessories that enable the Roto-Shake Genie, Enviro-Genie, Incubator Genie, and Rotator Genie to hold odd-shaped vessels.
Pack of 2 clip plates with two elastic bands each for use with odd-shaped sample vessels. This can be attached to the Magnetic Platform for use with the Roto-Shake Genie, Rotator Genie, Enviro-Genie or Incubator-Genie.
TRUSTED_SUSTAINABILITY
Specifications
Holds Odd-shaped sample vessels
Product Type Clip Plate
Material Stainless Steel
For Use With (Equipment) Roto-Shake Genie, Enviro-Genie, Incubator Genie, and Rotator Genie
Packaging Quantity 2 Clip Plates