Learn More
Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ High Performance Shakers
Product Code. 13127823 Shop All Thermo Scientific Products
Change view
Click to view available options
Product Type:
Test Tube Rack
Test Tube Rack Holder
Tube Size:
10 to 13 mm
16 to 20 mm
21 to 25 mm
26 to 30 mm
Varies, Adjustable
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Product Type Tube Size Unit Size
13127823 Test Tube Rack Holder Varies, Adjustable Each
10696173 Test Tube Rack 21 to 25 mm Each
10553503 Test Tube Rack 26 to 30 mm Each
13121144 Test Tube Rack 10 to 13 mm Each
10185612 Test Tube Rack 16 to 20 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 13127823 Supplier Thermo Scientific™ Supplier No. 600088
  • ONLINE EXCLUSIVE PRICE
    1005.00 €
    924.00 € / Pack of 1
    Save 81.00 €
    8% Off
Estimated Shipment: 30-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Shake multiple tubes at an adjustable angle to maximize sample aeration. The rack holders are adjustable to fit any of the four available test tube rack sizes.
Shake multiple tubes at an adjustable angle to maximize sample aeration. Thermo Scientific™ Universal Test Tube Racks with Holder for MaxQ™ 8000 Shakers provide ultimate flexibility—they can be adjusted to fit any of the four available test tube rack sizes.
  • Universal test tube rack holder can be adjusted to fit any of the four available test tube rack sizes
  • Universal design requires the separate purchase of a test tube rack
TRUSTED_SUSTAINABILITY
Specifications
Tube Size Varies, Adjustable
Product Type Test Tube Rack Holder
Description Universal adjustable-angle test tube rack holder
For Use With (Equipment) MaxQ HP 416, 420, 430, 435, 436, 480, 481, 8000