missing translation for 'onlineSavingsMsg'
Learn More
Learn More
uPAR Antibody (VIM-5), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11408-0.025mg
This item is not returnable.
View return policy
Description
uPAR Monoclonal antibody specifically detects uPAR in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| uPAR | |
| Monoclonal | |
| Unconjugated | |
| PBS | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| VIM-5 | |
| Flow Cytometry 1:50 - 1:200, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Frozen 1:100 | |
| CD87, CD87 antigen, Monocyte activation antigen Mo3, plasminogen activator, urokinase receptor, uPAR, U-PAR, UPARurokinase plasminogen activator surface receptor, u-plasminogen activator receptor form 2, URKRMO3 | |
| THP-1 cells (Human acute monocytic leukaemia cells). | |
| 0.025 mg | |
| Cancer | |
| 5329 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction