missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UQCRB Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
190.00 € - 470.00 €
Specifications
| Antigen | UQCRB |
|---|---|
| Dilution | Western Blot 1:1000-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18685830
|
Novus Biologicals
NBP2-93224-0.02ml |
0.02 mL |
190.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18681351
|
Novus Biologicals
NBP2-93224-0.1ml |
0.1 mL |
470.00 €
0.01mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
UQCRB Polyclonal antibody specifically detects UQCRB in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| UQCRB | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cardiovascular Biology, Cell Biology, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 7381 | |
| IgG | |
| Affinity purified |
| Western Blot 1:1000-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| Complex III subunit 7, Complex III subunit VII, cytochrome b-c1 complex subunit 7, QCR7, QPC, QP-CFLJ92016, ubiquinol-cytochrome c reductase binding protein, Ubiquinol-cytochrome c reductase complex 14 kDa protein, ubiquinol-cytochrome c reductase, complex III subunit VI, ubiquinone-binding protein, UQBC, UQBPFLJ97033, UQCR6, UQPC | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-111 of human UQCRB (NP_006285.1). MAGKQAVSASGKWLDGIRKWYYNAAGFNKLGLMRDDTIYEDEDVKEAIRRLPENLYNDRMFRIKRALDLNLKHQILPKEQWTKYEEENFYLEPYLKEVIRERKEREEWAKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title