Learn More
Invitrogen™ UQCRC2 Recombinant Rabbit Monoclonal Antibody (24GB2220)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557829
 View more versions of this product
Product Code. 30275572
  • 198.00 € / 20µL
Description
UQCRC2 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
This is a component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is part of the mitochondrial respiratory chain. The core protein 2 is required for the assembly of the complex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UQCRC2
Recombinant Monoclonal
Unconjugated
UQCRC2
1500004O06Rik; complex III subunit 2; Core protein II; cytochrome bc-1 complex core protein II; cytochrome b-c1 complex subunit 2, mitochondrial; MC3DN5; QCR2; U MC3DN5; ubiquinol cytochrome c reductase core protein 2; ubiquinol-cytochrome C reductase complex core protein 2, mitochondrial precursor (Complex III subunit II); ubiquinol-cytochrome c reductase core protein II; ubiquinol-cytochrome-c reductase complex core protein 2; UQCR2; Uqcrc2
Rabbit
Affinity chromatography
RUO
293448, 7385
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2220
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P22695, P32551
UQCRC2
A synthesized peptide derived from human UQCRC2 (aa 300-375).
20 μL
Primary
Human, Rat