Learn More
Invitrogen™ UQCRFS1 Recombinant Rabbit Monoclonal Antibody (24GB2460)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557831
 View more versions of this product
Product Code. 30275032
  • 198.00 € / 20µL
Description
UQCRFS1 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, Flow
Component of the ubiquinol-cytochrome c reductase complex (complex III or cytochrome b-c1 complex), which is a respiratory chain that generates an electrochemical potential coupled to ATP synthesis.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
UQCRFS1
Recombinant Monoclonal
Unconjugated
Uqcrfs1
4430402G14Rik; 8 kDa subunit 9; AI875505; Complex III subunit 5; Complex III subunit IX; Cytochrome b-c1 complex subunit 11; Cytochrome b-c1 complex subunit 5; Cytochrome b-c1 complex subunit 9; Cytochrome b-c1 complex subunit Rieske, mitochondrial; liver regeneration-related protein LRRGT00195; LRRGT00195; Rieske iron-sulfur protein; Rieske protein UQCRFS1; RIP1; RIS1; RISP; Su9; Ubiquinol-cytochrome c reductase 8 kDa protein; Ubiquinol-cytochrome c reductase iron-sulfur subunit; ubiquinol-cytochrome c reductase, Rieske iron-sulfur polypeptide 1; UQCR5; UQCRFS1
Rabbit
Affinity Chromatography
RUO
291103, 66694, 7386
-20°C
Liquid
Flow Cytometry, Western Blot, Immunocytochemistry
24GB2460
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P20788, P47985, Q9CR68
Uqcrfs1
A synthesized peptide derived from human UQCRFS1 (aa 220-274).
20 μL
Primary
Human, Mouse, Rat