missing translation for 'onlineSavingsMsg'
Learn More

Uranium Standard: U @ 1000 μg/mL in 5% HNO3, VHG™

Product Code. 17957989
Change view
Click to view available options
Quantity:
50 mL
100 mL
250 mL
500 mL
Unit Size:
100mL
250mL
500mL
50mL
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17957989 50 mL 50mL
17999269 100 mL 100mL
17948669 500 mL 500mL
17937679 250 mL 250mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 17957989 Supplier VHG Labs Supplier No. VHGPUN50
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

Uranium Standard: U @ 1000 μg/mL in 5% HNO3
TRUSTED_SUSTAINABILITY

Tekniske data

Chemical Name or Material Uranium
CAS 7440-61-1
Methods ICP/ICP-MS
Analyte or Component Names Uranium
Shipping Condition Room Temperature
Molecular Formula U
Certifications/Compliance ISO 17034
Synonym U
InChI Formula InChI=1S/U
SMILES [U]
IUPAC Name uranium
Solvent or Matrix Dilute HNO3
Molecular Weight (g/mol) 238.029
Quantity 50 mL
Formula Weight 238.051 g/mol
Vis mere Vis mindre
Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.