missing translation for 'onlineSavingsMsg'
Learn More

Ureidopropionic acid, MedChemExpress

Product Code. 16425576
Encompass_Preferred
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 16425576

Brand: Medchem Express HY113285

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Ureidopropionic acid (3-Ureidopropionic acid) is an intermediate in the metabolism of uracil.

Ureidopropionic acid (3-Ureidopropionic acid) is an intermediate in the metabolism of uracil.

Specifications

Chemical Name or Material Ureidopropionic acid
Physical Form Powder
CAS 462-88-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C4H8N2O3
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym 3-Ureidopropionic acid
Solubility Information DMSO : 125 mg/mL (946.11 mM; Need ultrasonic)
SMILES O=C(O)CCNC(N)=O
Molecular Weight (g/mol) 132.12
Formula Weight 132.12
Percent Purity 97.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.