Learn More
Invitrogen™ Urelumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA558416
 View more versions of this product
Product Code. 30284095
  • 1204.00 € / 1mg
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Urelumab has been used in trials studying the treatment of Leukemia, Multiple Myeloma, Malignant Tumors, and Cancer - Solid Tumors and B-Cell Non-Hodgkin's Lymphoma. Urelumab is a fully human antibody that targets CD137. The antibody product was developed using Medarex's UltiMAb(R) technology and was the first UltiMAb- derived antibody in clinical development by Bristol-Myers Squibb under the December 2003 agreement with Medarex.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Urelumab
Recombinant Monoclonal
Unconjugated
BMS-663513
Protein A
RUO
Human
Antibody
IgG4SP
ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance
1 mg/mL
25 mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2
Human
1 mg
Primary
-20°C, Avoid Freeze/Thaw Cycles
Lyophilized