missing translation for 'onlineSavingsMsg'
Learn More

Urolithin A, MedChemExpress

Product Code. 16492616
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492616 25 mg 25mg
16485827 5 mg 5mg
16453098 10 mg 10mg
16439656 50 mg 50mg
16434396 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16492616 Supplier Medchem Express Supplier No. HY100599
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Urolithin A, a gut-microbial metabolite of ellagic acid, exerts anti-inflammatory, antiproliferative, and antioxidant properties. Urolithin A induces autophagy and apoptosis, suppresses cell cycle progression, and inhibits DNA synthesis.

Urolithin A, a gut-microbial metabolite of ellagic acid, exerts anti-inflammatory, antiproliferative, and antioxidant properties. Urolithin A induces autophagy and apoptosis, suppresses cell cycle progression, and inhibits DNA synthesis.

Specifications

Chemical Name or Material Urolithin A
CAS 1143-70-0
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C13H8O4
Quantity 25 mg
Solubility Information DMSO : 30 mg/mL (131.46 mM; Need ultrasonic)
SMILES O=C1C2=CC(O)=CC=C2C3=CC=C(O)C=C3O1
Molecular Weight (g/mol) 228.2
Formula Weight 228.2
Percent Purity 95.57%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.