missing translation for 'onlineSavingsMsg'
Learn More

Urolithin B, TRC

Product Code. 30010413
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30010413 10 g 10g
30007572 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30010413 Supplier Toronto Research Chemicals Supplier No. TRCU84700510G
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1139-83-9
Molecular Formula C13H8O3
Molecular Weight (g/mol) 212.2
Synonym 3-Hydroxy-6H-dibenzo[b,d]pyran-6-one, 2',4'-Dihydroxy-2-biphenylcarboxylic Acid δ-Lactone, 3-Hydroxy-6H-benzo[c]chromen-6-one, 3-Hydroxydibenzo-α-pyrone, 3-Hydroxyurolithin, 7-Hydroxy-3,4-benzocoumarin, NSC 94726
SMILES Oc1ccc2c(OC(=O)c3ccccc23)c1

Specifications

Quantity 10 g
InChI Formula InChI=1S/C13H8O3/c14-8-5-6-10-9-3-1-2-4-11(9)13(15)16-12(10)7-8/h1-7,14H
Formula Weight 212.0473
Percent Purity >95%
Chemical Name or Material Urolithin B
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.