missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ia Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
193.00 € - 468.00 €
Specifications
| Antigen | Uroplakin Ia |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18673541
|
Novus Biologicals
NBP2-93229-0.02ml |
0.02 mL |
193.00 €
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18697791
|
Novus Biologicals
NBP2-93229-0.1ml |
0.1 mL | |||||||
Description
Uroplakin Ia Polyclonal antibody specifically detects Uroplakin Ia in Mouse, Rat samples. It is validated for Western BlotSpecifications
| Uroplakin Ia | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 11045 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Mouse, Rat | |
| TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 110-230 of human UPK1A (NP_008931.1). SCITSYTHRDYMVSNPSLITKQMLTFYSADTDQGQELTRLWDRVMIEQECCGTSGPMDWVNFTSAFRAATPEVVFPWPPLCCRRTGNFIPLNEEGCRLGHMDYLFTKGCFEHIGHAIDSYT | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title