missing translation for 'onlineSavingsMsg'
Learn More

Uroplakin Ia, Mouse anti-Human, Clone: UPK1A/2925, Novus Biologicals™

Mouse Monoclonal Antibody

281.00 € - 512.00 €

Specifications

Antigen Uroplakin Ia
Clone UPK1A/2925
Concentration 0.2 mg/mL
Dilution ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Applications ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18215326
View Documents
Novus Biologicals
NBP2-79735-20UG
20 μg
281.00 €
20µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18238137
View Documents
Novus Biologicals
NBP2-79735-100UG
100 μg
512.00 €
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Uroplakin Ia Monoclonal specifically detects Uroplakin Ia in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications

Specifications

Uroplakin Ia
0.2 mg/mL
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array
Unconjugated
Mouse
Human
TSPAN21, UP1A, UPIA, UPK1A uroplakin 1A, UPKA
UPK1A
IgG1 κ
Protein A or G purified
UPK1A/2925
ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array
Monoclonal
Purified
RUO
10 mM PBS with 0.05% BSA
11045
Recombinant fragment (around aa 114-173) of human Uroplakin Ia protein (exact sequence is proprietary)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.