missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HCV-NS5b Antibody (5B-12B7), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-80124JF549
This item is not returnable.
View return policy
Description
HCV-NS5b Monoclonal antibody specifically detects HCV-NS5b in Virus samples. It is validated for Immunofluorescence
Specifications
| HCV-NS5b | |
| Monoclonal | |
| Janelia Fluor 549 | |
| EC:2.7.7.48, Nonstructural Protein 5B, NS5B, Q8JXY1, RNA-directed RNA polymerase | |
| Recombinant HCV-NS5b RNA-dependent RNA polymerase. | |
| 0.1 mL | |
| Virology Bacteria and Parasites | |
| 951475 | |
| Store at 4°C in the dark. | |
| IgG2a |
| Immunofluorescence | |
| 5B-12B7 | |
| 50mM Sodium Borate | |
| Mouse | |
| >95%, by SDS-PAGE | |
| RUO | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction