missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Uroplakin Ib, Mouse anti-Human, Clone: UPK1B/3273, Novus Biologicals™
Mouse Monoclonal Antibody
268.00 € - 489.00 €
Specifications
| Antigen | Uroplakin Ib |
|---|---|
| Clone | UPK1B/3273 |
| Concentration | 0.2 mg/mL |
| Dilution | ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array |
| Applications | ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18208887
|
Novus Biologicals
NBP2-79801-20UG |
20 μg |
268.00 €
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18212077
|
Novus Biologicals
NBP2-79801-100UG |
100 μg |
489.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Uroplakin Ib Monoclonal specifically detects Uroplakin Ib in Human samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.Specifications
| Uroplakin Ib | |
| 0.2 mg/mL | |
| ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| Unconjugated | |
| Mouse | |
| Human | |
| tetraspanin-20, Tspan-20, TSPAN20tetraspan, UP1b, UPIb, UPK1, uroplakin 1B, Uroplakin Ib, uroplakin-1b | |
| UPK1B | |
| IgG1 κ | |
| Protein A or G purified |
| UPK1B/3273 | |
| ELISA, Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| Monoclonal | |
| Purified | |
| RUO | |
| 10 mM PBS with 0.05% BSA | |
| 7348 | |
| Recombinant fragment (around aa 109-229) of human Uroplakin Ib protein (exact sequence is proprietary) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title