Learn More
Memmert™ USB User-ID Stick (with User-ID Licence) for SN75plus
Product Code. 17598775 Shop All Memmert Products
Change view
Click to view available options
For Use With (Equipment):
SN75plus Sterilizer
Unit Size:
Each
Product Code. For Use With (Equipment) Unit Size
17598775 SN75plus Sterilizer Each
1 options
Product Code. 17598775 Supplier Memmert™ Supplier No. SN75plusB33170
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
USB User-ID Stick prevents undesired manipulation by unauthorized third parties.
  • Oven-linked authorization licence (User-ID-program) on Memory-stick
  • Prevents undesired manipulation by unauthorised third parties
  • When reordering please specify serial number (only for units with TwinDISPLAY)
Specifications
For Use With (Equipment) SN75plus Sterilizer
Product Type USB Stick
Includes User ID Licence