Learn More
USF1 Antibody (S03-5E6), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP3-19925-50ul
 View more versions of this product
Product Code. 18668344
  • 421.00 € / 50µL
Description
USF1 Monoclonal antibody specifically detects USF1 in Human, Mouse, Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
USF1
Monoclonal
0.3 mg/mL
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Western Blot
S03-5E6
Unconjugated
BHLHB11, bHLHb11FCHL1, Class B basic helix-loop-helix protein 11, FCHL, HYPLIP1, Major late transcription factor 1, MLTF, MLTFI, UEF, upstream stimulatory factor 1, upstream transcription factor 1, USF
A synthetic peptide of human USF1 (Uniprot # P22415)
50 μg
Transcription Factors and Regulators
7391
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.