missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP10 Monoclonal Antibody (OTI1H9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504007S
This item is not returnable.
View return policy
Description
USP10 Monoclonal antibody specifically detects USP10 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| USP10 | |
| Monoclonal | |
| 0.74 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P23743 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1606 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H9 | |
| Unconjugated | |
| Dgka | |
| 2610014N07Rik; Deubiquitinating enzyme 10; KIAA0190; MGC2621; mKIAA0190; Ode-1; ubiquintin c-terminal hydrolase related polypeptide; ubiquitin carboxyl-terminal hydrolase 10; ubiquitin specific peptidase 10; ubiquitin specific protease 10; ubiquitin thioesterase 10; ubiquitin thiolesterase 10; ubiquitin-specific-processing protease 10; UBPO; Uchrp; USP10 | |
| Full length human recombinant protein of human DGKA produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction