missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP14 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 539.00 €
Specifications
| Antigen | USP14 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18465201
|
Novus Biologicals
NBP1-86946-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18281688
|
Novus Biologicals
NBP1-86946 |
0.1 mL |
539.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP14 Polyclonal specifically detects USP14 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| USP14 | |
| Polyclonal | |
| Rabbit | |
| Protein Turnover, Ubiquitin Proteasome Pathway | |
| P54578 | |
| 9097 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:EKESVNAKVLKDVKFPLMLDMYELCTPELQEKMVSFRSKFKDLEDKKVNQQPNTSDKKSSPQKEVKYEPFSFADDIGSNNCGYYDLQAVLTHQGRSSSSGHYVSWVKRKQDEWIKFDDDKVSIVTPEDILRLSGGGDWHIAYVL | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| 56 kDa |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:500 - 1:1000, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:500-1:1000 | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| Deubiquitinating enzyme 14, EC 3.1.2.15, EC 3.4.19.12, TGT, TGTubiquitin carboxyl-terminal hydrolase 14, tRNA-guanine transglycosylase, 60-kD subunit, ubiquitin specific peptidase 14 (tRNA-guanine transglycosylase), ubiquitin specific protease 14 (tRNA-guanine transglycosylase), ubiquitin thioesterase 14, Ubiquitin thiolesterase 14, ubiquitin-specific processing protease 14, Ubiquitin-specific-processing protease 14 | |
| USP14 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title