missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP14 Rabbit Recombinant Antibody, Proteintech
Rabbit Recombinant Antibody
129.00 € - 399.00 €
Specifications
| Antigen | USP14 |
|---|---|
| Clone | 3G12 |
| Concentration | 400 μg/mL |
| Applications | Western Blot, Immunoprecipitation, ELISA |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30236427
|
Proteintech
82128-1-RR-20UL |
20 μL |
149.00 € 129.00 € / 20µL Save 20.00 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30233434
|
Proteintech
82128-1-RR-100UL |
100 μL |
439.00 € 399.00 € / 100µL Save 40.00 € 9% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The USP14 antibody from Proteintech is a rabbit recombinant antibody generated with recombinant protein of human USP14. This antibody recognizes human, mouse, rat antigen. The USP14 antibody has been validated for the following applications: WB, IP, ELISA analysis.Specifications
| USP14 | |
| 400 μg/mL | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| 9097 | |
| Recombinant Protein | |
| Primary | |
| Store at -20°C. Stable for one year after shipment. |
| 3G12 | |
| Western Blot, Immunoprecipitation, ELISA | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| TGT | |
| IgG | |
| Protein A purification | |
| Recombinant Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title