missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP19 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93529-0.1ml
This item is not returnable.
View return policy
Description
USP19 Polyclonal antibody specifically detects USP19 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| USP19 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| Deubiquitinating enzyme 19, EC 3.1.2.15, EC 3.4.19.12, KIAA0891, ubiquitin carboxyl-terminal hydrolase 19, ubiquitin specific peptidase 19, ubiquitin thioesterase 19, Ubiquitin thiolesterase 19, Ubiquitin-specific-processing protease 19, Zinc finger MYND domain-containing protein 9, ZMYND9ubiquitin specific protease 19 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human USP19 (NP_001186089.1). MSGGASATGPRRGPPGLEDTTSKKKQKDRANQESKDGDPRKETGSRYVAQAGLEPLASGDPSASASHAAGITGSRHRTRLFFPSSSGSASTPQEEQTKEGACEDPHDLLATPTPELLLDWRQSAEEVIVKLRVGVGPLQLEDVDAAFTDTDCVVRFAGGQQWGGVFYAEIKSSCAKVQTRKGSLLHLTLPKKVPMLTWPS | |
| 0.1 mL | |
| Neuroscience | |
| 10869 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction