missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP40 Antibody (OTI8F8), Novus Biologicals Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-45036-100ul
This item is not returnable.
View return policy
Description
USP40 Monoclonal antibody specifically detects USP40 in Human samples. It is validated for Western Blot
Specifications
| USP40 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.3) 1% BSA, 50% glycerol | |
| Mouse | |
| >95% | |
| RUO | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI8F8 | |
| Western Blot 1:500-2000 | |
| deubiquitinating enzyme 40, ubiquitin specific peptidase 40, ubiquitin thiolesterase 40, ubiquitin-specific-processing protease 40 | |
| Human recombinant protein fragment corresponding to amino acids 767-912 of human USP40(NP_060688) produced in E.coli. | |
| 100 μL | |
| Cell Biology | |
| 55230 | |
| Store at -20 degrees C. Avoid freeze/thaw cycles. | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction