missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP46 Antibody (CL0363) [mFluor Violet 610 SE], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-52923MFV610
This item is not returnable.
View return policy
Description
USP46 Monoclonal antibody specifically detects USP46 in Human samples. It is validated for Western Blot,KnockDown,Immunocytochemistry/ Immunofluorescence
Specifications
| USP46 | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| Deubiquitinating enzyme 46, EC 3.1.2.15, EC 3.4.19.12, FLJ11850, FLJ12552, FLJ14283, FLJ39393, ubiquitin carboxyl-terminal hydrolase 46, ubiquitin specific peptidase 46, ubiquitin specific protease 46, ubiquitin thioesterase 46, Ubiquitin thiolesterase 46, Ubiquitin-specific-processing protease 46 | |
| This antibody was developed using a recombinant protein derived from P62068, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 64854 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Western Blot, KnockDown, Immunocytochemistry/Immunofluorescence | |
| CL0363 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction