missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP6NL Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93558-0.1ml
This item is not returnable.
View return policy
Description
USP6NL Polyclonal antibody specifically detects USP6NL in Human samples. It is validated for Western Blot
Specifications
| USP6NL | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| KIAA0019RNTRErelated to the N terminus of tre, Related to the N-terminus of tre, RN-tre, TRE2NL, USP6 N-terminal like, USP6 N-terminal-like protein | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 300-600 of human USP6NL (NP_055503.1). SYTILKLHKKHLMKLSMEELVEFFQETLAKDFFFEDDFVIEQLQISMTELKRAKLDLPEPGKEDEYPKKPLGQLPPELQSWGVHHLSNGQRSVGRPSPLASGRRESGAPHRRHEHSPHPQSRTGTPERAQPPRRKSVEEESKKLKDEADFQRKLPSGPQDSSRQYNHAAANQNSNATSNIRKEFVPKWNKPSDVSATERTAKYTMEGKGRAAHPALAVTVPGPAEVRVSNVRPKMKALDAEDGKRGSTASQYDNVPGPELDSGASVEEALERAYSQSPRHALYPPSPRKHAEPSSSPSKVS | |
| 0.1 mL | |
| GPCR, Signal Transduction | |
| 9712 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction