missing translation for 'onlineSavingsMsg'
Learn More
Learn More
USP7 Antibody (06), Novus Biologicals™
Mouse Monoclonal Antibody
220.00 € - 326.00 €
Specifications
| Antigen | USP7 |
|---|---|
| Clone | 06 |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18661328
|
Novus Biologicals
NBP3-06428-100ul |
100 μg |
326.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18649277
|
Novus Biologicals
NBP3-06428-50ul |
50 μg |
220.00 €
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
USP7 Monoclonal antibody specifically detects USP7 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| USP7 | |
| Immunohistochemistry-Paraffin 1:50-1:200 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Deubiquitinating enzyme 7, EC 3.1.2.15, EC 3.4.19.12, HAUSPTEF1, Herpes virus-associated ubiquitin-specific protease, Herpesvirus-associated ubiquitin-specific protease, ubiquitin carboxyl-terminal hydrolase 7, ubiquitin specific peptidase 7 (herpes virus-associated), ubiquitin specific protease 7 (herpes virus-associated), Ubiquitin thioesterase 7, Ubiquitin-specific-processing protease 7 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human USP7 (Accession#: NP_003461.2; Lys208-Glu560). | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 06 | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Cell Biology, DNA Repair, Neuroscience, Stem Cell Markers, Ubiquitin Proteasome Pathway | |
| 0.2 um filtered solution in PBS | |
| 7874 | |
| IgG2a | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title