missing translation for 'onlineSavingsMsg'
Learn More

Greiner Bio-One UV-Star™ 96-well UV Spectroscopy Microplate

Product Code. 10781244
Change view
Click to view available options
Material:
Cycloolefin Copolymer
Polystyrene
Well Design:
Chimney
Half area
Unit Size:
Pack of 40
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Well Design Unit Size
10781244 Polystyrene Half area Pack of 40
10600942 Cycloolefin Copolymer Chimney Pack of 40
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10781244 Supplier Greiner Bio-One Supplier No. 675801
  • ONLINE EXCLUSIVE PRICE
    595.00 €
    439.61 € / Pack of 40
    Save 155.39 €
    26% Off
View all promotions
Estimated Shipment: 11-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use in UV Spectroscopy. Greiner Bio-One UV-Star™ 96-Well UV Spectroscopy Microplate is used for measurements of protein concentration at 280nm.

  • Free of detectable DNase, RNase, human DNA
  • Non-pyrogenic
  • Manufactured under DIN ISO 9001 guidelines
  • Can be traced back to production through defined LOT number system
  • Optical window down to 230nm ideal for nucleic acid

Service Plan

No

Warranty

No

TRUSTED_SUSTAINABILITY

Specifications

Barcode Alphanumeric
Breakapart No
Format Half-Area
Array 8 x 12
Packaging Inner bags of 10
Binding Type Non Binding
Color Clear
Product Type UV Microplate
Material Polystyrene
Volume (Metric) Well 15 to 175 μL
No. of Wells 96
Sterility Non-sterile
Surface Treatment Untreated
Total Volume (Metric) Well 199 μL
Well Shape Flat Bottom (F-well)
Dnase Free Yes
For Use With (Application) UV Spectroscopy
Quantity 40/Cs.
Well Design Half area
Dimensions (L x W x H) 127.8 x 85.5 x 11.5 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.