missing translation for 'onlineSavingsMsg'
Learn More
Learn More
UVSSA Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06046-50ul
This item is not returnable.
View return policy
Description
UVSSA Polyclonal antibody specifically detects UVSSA in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| UVSSA | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| hypothetical protein LOC57654, KIAA1530, MGC117169 | |
| Produced in rabbits immunized with E. coli-derived Human UVSSA fragment. | |
| 50 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 57654 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur