missing translation for 'onlineSavingsMsg'
Learn More

V21+K Wear Metals Standard: 100 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG™

Product Code. 16499669
Change view
Click to view available options
Quantity:
100 g
200 g
400 g
Unit Size:
100g
200g
400g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16499669 100 g 100g
17960956 400 g 400g
17922926 200 g 200g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16499669 Supplier VHG Labs Supplier No. VHGV21+K100100G
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

V21+K Wear Metals Standard: 100 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
CAS 7429-90-5, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2, 7440-47-3
Methods ICP/XRF/RDE
Analyte or Component Names Aluminium/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hydrocarbon Oil
Quantity 100 g
Concentration or Composition (by Analyte or Components) Aluminium 100 μg/g/Barium 100 μg/g/Boron 100 μg/g/Cadmium 100 μg/g/Calcium 100 μg/g/Chromium 100 μg/g/Copper 100 μg/g/Iron 100 μg/g/Lead 100 μg/g/Magnesium 100 μg/g/Manganese 100 μg/g/Molybdenum 100 μg/g/Nickel 100 μg/g/Phosphorus 100 μg/g/Potassium 100 μg/g/Silicon 100 μg/g/Silver 100 μg/g/Sodium 100 μg/g/Tin 100 μg/g/Titanium 100 μg/g/Vanadium 100 μg/g/Zinc 100 μg/g
compliance-icons
Product Identifier
  • Conostan S21 Element Blend
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contain: White mineral oil, petroleum
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.