missing translation for 'onlineSavingsMsg'
Learn More

V23 Wear Metals Standard: 500 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil, VHG™

Product Code. 17951996
Change view
Click to view available options
Quantity:
50 g
100 g
200 g
400 g
Unit Size:
100g
200g
400g
50g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17951996 200 g 200g
17970636 400 g 400g
17921856 100 g 100g
17984026 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 17951996 Supplier VHG Labs Supplier No. VHGV23500200G
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

VHG Labs offers ISO 17034 metallo-organic and aqueous inorganic certified reference materials for oil, fuel, and coolant analysis as well as a variety of certified standards for environmental, pharmaceutical, and industrial applications

V23 Wear Metals Standard: 500 μg/g Ag, Al, B, Ba, Ca, Cd, Cr, Cu, Fe, K, Mg, Mn, Mo, Na, Ni, P, Pb, Sb, Si, Sn, Ti, V, Zn in Hydrocarbon Oil
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aluminium/Antimony/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
CAS 7429-90-5, 7440-36-0, 7440-39-3, 7440-42-8, 7440-43-9, 7440-70-2
Methods ICP/XRF/RDE
Analyte or Component Names Aluminium/Antimony/Barium/Boron/Cadmium/Calcium/Chromium/Copper/Iron/Lead/Magnesium/Manganese/Molybdenum/Nickel/Phosphorus/Potassium/Silicon/Silver/Sodium/Tin/Titanium/Vanadium/Zinc
Shipping Condition Room Temperature
Certifications/Compliance ISO 17034
Solvent or Matrix Hydrocarbon Oil
Quantity 200 g
Concentration or Composition (by Analyte or Components) Aluminium 500 μg/g/Antimony 500 μg/g/Barium 500 μg/g/Boron 500 μg/g/Cadmium 500 μg/g/Calcium 500 μg/g/Chromium 500 μg/g/Copper 500 μg/g/Iron 500 μg/g/Lead 500 μg/g/Magnesium 500 μg/g/Manganese 500 μg/g/Molybdenum 500 μg/g/Nickel 500 μg/g/Phosphorus 500 μg/g/Potassium 500 μg/g/Silicon 500 μg/g/Silver 500 μg/g/Sodium 500 μg/g/Tin 500 μg/g/Titanium 500 μg/g/Vanadium 500 μg/g/Zinc 500 μg/g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.