missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VAC14 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17844450
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17844450 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17844450 Supplier Invitrogen™ Supplier No. PA5139933

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: IECPIFTYLR LQLLDVKNNP YLIKALYGLL MLLPQSSAFQ LLSHRLQCVP NPELLQTEDS LKAAPKSQKA DSPSIDYAEL LQHFEKVQNK HLEV Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 94%.

Vac14 a putative cytoskeletal interacting protein; mutation of gene may be associated with lethal communicating hydrocephalus with perinatal onset.
TRUSTED_SUSTAINABILITY

Specifications

Antigen VAC14
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.4 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Vac14
Gene Accession No. Q08AM6
Gene Alias AA959718; ArPIKfyve; BC032215; D8Wsu151e; ingls; Protein VAC14 homolog; Tax1 (human T-cell leukemia virus type I) binding protein 1; Tax1 (human T-cell leukemia virus type I) binding protein 2; tax1-binding protein 2; TAX1BP2; TRX; Vac14; Vac14 homolog; Vac14 homolog (S. cerevisiae); Vac14, PIKFYVE complex component; vacuole 14 protein; vacuole morphology 14
Gene Symbols Vac14
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human VAC14. Recombinant protein control fragment (Product #RP-109476).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55697
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.