missing translation for 'onlineSavingsMsg'
Learn More

Vadimezan, MedChemExpress

Product Code. 16484318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16484318 10 mg 10mg
16477037 5 mg 5mg
16440647 50 mg 50mg
16445236 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16484318 Supplier Medchem Express Supplier No. HY10964
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Vadimezan (DMXAA; ASA-404), the tumor vascular disrupting agent (tumor-VDA), is a murine agonist of the stimulator of interferon genes (STING) and also a potent inducer of type I IFNs and other cytokines. Vadimezan has anti-influenza virus H1N1-PR8 activities.

Vadimezan (DMXAA

Specifications

Chemical Name or Material Vadimezan
CAS 117570-53-3
Color White
Purity Grade Notes Research
Molecular Formula C17H14O4
Quantity 10 mg
Synonym DMXAA ASA-404
Solubility Information DMSO : 7.14 mg/mL (25.29 mM; Need ultrasonic) ∣7.5% sodium bicarbonate : 6.67 mg/mL (23.63 mM; Need ultrasonic)
SMILES O=C(O)CC1=CC=CC(C2=O)=C1OC3=C2C=CC(C)=C3C
Molecular Weight (g/mol) 282.29
Formula Weight 282.29
Percent Purity 99.44%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.