missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Valproic Acid Monoclonal Antibody (VP13-615.3), Invitrogen™
Mouse Monoclonal Antibody
424.00 € - 1052.00 €
Specifications
| Antigen | Valproic Acid |
|---|---|
| Clone | VP13-615.3 |
| Concentration | 2 mg/mL |
| Applications | ELISA |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17830486
|
Invitrogen™
MA5-46043 |
100 μg |
424.00 €
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
11549032
|
Invitrogen
MA110769 |
500 μg |
538.00 €
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17850176
|
Invitrogen™
MA5-46141 |
1 mg |
1052.00 €
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MA1-10769 detects Valproic Acid. MA1-10769 has been successfully used in ELISA applications. The MA1-10769 immunogen is Valproic acid, conjugated to KLH.
Valproic acid, valproate sodium, and divalproex belong to the group of medicines called anticonvulsants. They are used to control certain types of seizures in the treatment of epilepsy. Also Valproic acid relieves histone deacetylases (HDACs) dependent transcriptional repression and causes hyperacetylation of histones in cultured cells and in vivo. Valproic acid inhibits HDAC activity in vitro, most probably by binding to the catalytic center of HDACs. Most importantly, valproic acid induces differentiation of carcinoma cells, transformed hematopoietic progenitor cells and leukemic blasts from acute myeloid leukemia patients. More over, tumor growth and metastasis formation are significantly reduced in animal experiments. Therefore, valproic acid might serve as an effective drug for cancer therapy.Specifications
| Valproic Acid | |
| 2 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with no preservative; pH 7.4 | |
| Valproic acid, conjugated to KLH. | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles |
| VP13-615.3 | |
| ELISA | |
| Unconjugated | |
| Mouse | |
| Chemical | |
| C8H16O2; Depakene; Depakine; Dipropylacetic acid | |
| IgG1 | |
| Protein A | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title