missing translation for 'onlineSavingsMsg'
Learn More

Valproic acid sodium, MedChemExpress

Product Code. 16406767
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16406767 5 g 5g
16472936 25 g 25g
16421069 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16406767 Supplier Medchem Express Supplier No. HY10585A
 Restricted Item
Estimated Shipment: 08-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Valproic acid (Sodium Valproate) sodium is an orally active HDAC inhibitor, with IC50 in the range of 0.5 and 2 mM, also inhibits HDAC1 (IC50, 400 μM), and induces proteasomal degradation of HDAC2.

Valproic acid sodium activates Notch1 signaling and inhibits proliferation in small cell lung cancer (SCLC) cells. Valproic acid sodium is used in the treatment of epilepsy, bipolar disorder, metabolic disease, HIV infection and prevention of migraine headaches.

Specifications

Chemical Name or Material Valproic acid sodium
Physical Form Solid
CAS 1069-66-5
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C8H15NaO2
For Use With (Application) Cancer-Kinase/protease
Synonym Sodium Valproate sodium
Solubility Information H2O : 250 mg/mL (1504.30 mM; Need ultrasonic)
SMILES O=C(O[Na])C(CCC)CCC
Molecular Weight (g/mol) 166.19
Formula Weight 166.19
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.