missing translation for 'onlineSavingsMsg'
Learn More

Valspodar, MedChemExpress

Product Code. 16477318
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16477318 10 mg 10mg
16419827 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16477318 Supplier Medchem Express Supplier No. HY17384
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Valspodar (PSC 833) is a selective P-glycoprotein inhibitor that has been used as an experimental cancer treatment and chemosensitizer.

Valspodar (PSC 833) is a selective P-glycoprotein inhibitor that has been used as an experimental cancer treatment and chemosensitizer.

Specifications

Chemical Name or Material Valspodar
Physical Form Solid
CAS 121584-18-7
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C63H111N11O12
For Use With (Application) Cancer-programmed cell death
Synonym PSC 833
Solubility Information DMSO : 12 mg/mL (9.88 mM; Need ultrasonic and warming)
SMILES O=C([C@H](C(C)C)N(C)C([C@H](CC(C)C)N(C)C([C@H](CC(C)C)N(C)C([C@@H](C)NC([C@H](C)NC([C@H](CC(C)C)N(C)C([C@H](C(C)C)NC([C@H](CC(C)C)N(C)C(CN(C)C([C@H](C(C)C)N1)=O)=O)=O)=O)=O)=O)=O)=O)=O)N(C)[C@@H](C([C@H](C)C/C=C/C)=O)C1=O
Molecular Weight (g/mol) 1214.62
Formula Weight 1214.62
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.