missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ VAMP1/VAMP2 Monoclonal Antibody (SP10)

Mouse Monoclonal Antibody

Brand:  Invitrogen™ MA517608

Product Code. 15368510

  • 520.00 € / 200µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MA5-17608 targets VAMP/synaptobrevin in ELISA, FACS, and WB applications and shows reactivity with Human samples. The MA5-17608 immunogen is human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).

Vesicle-associated membrane protein 2 is essential for cAMP-regulated exocytosis. VAMP (also know as synaptobrevin) is part of the synaptic vesicle docking and fusion complex and plays a central role in neuroexocytosis. Two VAMP (vesicle-associated membrane protein) isoforms are expressed in the nervous system and are differently distributed among the specialized parts of the tissue. VAMP-1 and -2 are present in all tissues tested, including kidney, adrenal gland, liver, pancreas, thyroid, heart, and smooth muscle. The two isoforms are differentially expressed in various tissues and their level may depend on differentiation. VAMP-1 is restricted to exocrine pancreas and to kidney tubular cells, whereas VAMP-2 is the predominant isoform present in Langerhans islets and in glomerular cells. Both isoforms show a patchy vesicular intracellular distribution in confocal microscopy. Evidence for the importance of neuronal VAMPproteins in the physiology of all cells is indicated.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

VAMP1/VAMP2
Monoclonal
1 mg/mL
PBS with 0.02% sodium azide
P23763, P63027
VAMP1, VAMP2
Human Synaptobrevin 1 (aa 1-118) and Synaptobrevin 2(aa 33-96).
200 μg
Primary
Human
Antibody
IgM
ELISA, Flow Cytometry, Western Blot
SP10
Unconjugated
VAMP1
RATVAMPB; RATVAMPIR; SPAX1; SYB; SYB1; SYB2; synaptobrevin 1; synaptobrevin 2; Synaptobrevin 2 (vesicle-associated membrane protein VAMP-2); synaptobrevin-1; Synaptobrevin-2; Vamp1; VAMP-1; VAMP2; VAMP-2; vesicle associated membrane protein 1; vesicle associated membrane protein 2; Vesicle-associated membrane protein (synaptobrevin 2); vesicle-associated membrane protein 1; vesicle-associated membrane protein 1 (synaptobrevin 1); vesicle-associated membrane protein 2; vesicle-associated membrane protein 2 (synaptobrevin 2)
Mouse
Affinity chromatography
RUO
6843, 6844
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.